Merck
  • Plasmin inhibitors prevent leukocyte accumulation and remodeling events in the postischemic microvasculature.
Plasmin inhibitors prevent leukocyte accumulation and remodeling events in the postischemic microvasculature.
PloS one (2011-03-03)
Christoph A Reichel, Max Lerchenberger, Bernd Uhl, Markus Rehberg, Nina Berberich, Stefan Zahler, Matthias P Wymann, Fritz Krombach
ZUSAMMENFASSUNG
Clinical trials revealed beneficial effects of the broad-spectrum serine protease inhibitor aprotinin on the prevention of ischemia-reperfusion (I/R) injury. The underlying mechanisms remained largely unclear. Using in vivo microscopy on the cremaster muscle of male C57BL/6 mice, aprotinin as well as inhibitors of the serine protease plasmin including tranexamic acid and ε-aminocaproic acid were found to significantly diminish I/R-elicited intravascular firm adherence and (subsequent) transmigration of neutrophils. Remodeling of collagen IV within the postischemic perivenular basement membrane was almost completely abrogated in animals treated with plasmin inhibitors or aprotinin. In separate experiments, incubation with plasmin did not directly activate neutrophils. Extravascular, but not intravascular administration of plasmin caused a dose-dependent increase in numbers of firmly adherent and transmigrated neutrophils. Blockade of mast cell activation as well as inhibition of leukotriene synthesis or antagonism of the platelet-activating-factor receptor significantly reduced plasmin-dependent neutrophil responses. In conclusion, our data suggest that extravasated plasmin(ogen) mediates neutrophil recruitment in vivo via activation of perivascular mast cells and secondary generation of lipid mediators. Aprotinin as well as the plasmin inhibitors tranexamic acid and ε-aminocaproic acid interfere with this inflammatory cascade and effectively prevent postischemic neutrophil responses as well as remodeling events within the vessel wall.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ginkgolid B aus Ginkgobiloba-Blätter, ≥90% (HPLC)