Merck
  • The mRNA-edited form of GABRA3 suppresses GABRA3-mediated Akt activation and breast cancer metastasis.
The mRNA-edited form of GABRA3 suppresses GABRA3-mediated Akt activation and breast cancer metastasis.
Nature communications (2016-02-13)
Kiranmai Gumireddy, Anping Li, Andrew V Kossenkov, Masayuki Sakurai, Jinchun Yan, Yan Li, Hua Xu, Jian Wang, Paul J Zhang, Lin Zhang, Louise C Showe, Kazuko Nishikura, Qihong Huang
ZUSAMMENFASSUNG
Metastasis is a critical event affecting breast cancer patient survival. To identify molecules contributing to the metastatic process, we analysed The Cancer Genome Atlas (TCGA) breast cancer data and identified 41 genes whose expression is inversely correlated with survival. Here we show that GABAA receptor alpha3 (Gabra3), normally exclusively expressed in adult brain, is also expressed in breast cancer, with high expression of Gabra3 being inversely correlated with breast cancer survival. We demonstrate that Gabra3 activates the AKT pathway to promote breast cancer cell migration, invasion and metastasis. Importantly, we find an A-to-I RNA-edited form of Gabra3 only in non-invasive breast cancers and show that edited Gabra3 suppresses breast cancer cell invasion and metastasis. A-to-I-edited Gabra3 has reduced cell surface expression and suppresses the activation of AKT required for cell migration and invasion. Our study demonstrates a significant role for mRNA-edited Gabra3 in breast cancer metastasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human GABRA3