Merck
  • Up-regulated miR-145 expression inhibits porcine preadipocytes differentiation by targeting IRS1.
Up-regulated miR-145 expression inhibits porcine preadipocytes differentiation by targeting IRS1.
International journal of biological sciences (2012-12-01)
Yunxue Guo, Yaosheng Chen, Yun Zhang, Yue Zhang, Luxi Chen, Delin Mo
ZUSAMMENFASSUNG
Generally, most miRNAs that were up-regulated during differentiation promoted adipogenesis, but our research indicated that up-regulation of miR-145 in porcine preadipocytes did not promote but inhibit adipogenesis. In this study, miR-145 was significantly up-regulated during porcine dedifferentiated fat (DFAT) cells differentiation. In miR-145 overexpressed DFAT cells, adipogenesis was inhibited and triglycerides accumulation was decreased after hormone stimulation (P<0.05). Furthermore, up-regulation of miR-145 expression repressed induction of mRNA levels of adipogenic markers, such as CCAAT/enhancer-binding protein α (C/EBPα), and peroxisome proliferator-activated receptor γ2 (PPARγ2). These effects caused by miR-145 overexpression were mediated by Insulin receptor substrate 1 (IRS1) as a mechanism. These data suggested that induced miR-145 expression during differentiation could inhibit adipogenesis by targeting IRS1, and miR-145 may be novel agent for adipose tissue engineering.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-IRS1-Antikörper, Klon 4.2.2, clone 4.2.2, from mouse