Merck
  • Cytosolic aminopeptidases influence MHC class I-mediated antigen presentation in an allele-dependent manner.
Cytosolic aminopeptidases influence MHC class I-mediated antigen presentation in an allele-dependent manner.
Journal of immunology (Baltimore, Md. : 1950) (2009-11-18)
Eunkyung Kim, Heechun Kwak, Kwangseog Ahn
ZUSAMMENFASSUNG
Antigenic peptides presented by MHC class I molecules are generated mainly by the proteasome in the cytosol. Several cytosolic aminopeptidases further trim proteasomal products to form mature epitopes or individual amino acids. However, the distinct function of cytosolic aminopeptidases in MHC class I Ag processing remains to be elucidated. In this study, we show that cytosolic aminopeptidases differentially affect the cell surface expression of MHC class I molecules in an allele-dependent manner in human cells. In HeLa cells, knockdown of puromycin-sensitive aminopeptidase (PSA) by RNA interference inhibited optimal peptide loading of MHC class I molecules, and their cell surface expression was correspondingly reduced. In contrast, depletion of bleomycin hydrolase (BH) enhanced optimal peptide loading and cell surface expression of MHC class I molecules. We did not find evidence on the effect of leucine aminopeptidase knockdown on the MHC class I Ag presentation. Moreover, we demonstrated that PSA and BH influence the peptide loading and surface expression of MHC class I in an allele-specific manner. In the absence of either PSA or BH, the surface expression and peptide-dependent stability of HLA-A68 were reduced, whereas those of HLA-B15 were enhanced. The surface expression and peptide-dependent stability of HLA-A3 were enhanced by BH knockdown, although those of HLA-B8 were increased in PSA-depleted conditions.