Merck
  • A mechanism for translationally coupled mRNA turnover: interaction between the poly(A) tail and a c-fos RNA coding determinant via a protein complex.
A mechanism for translationally coupled mRNA turnover: interaction between the poly(A) tail and a c-fos RNA coding determinant via a protein complex.
Cell (2000-10-29)
C Grosset, C Y Chen, N Xu, N Sonenberg, H Jacquemin-Sablon, A B Shyu
ZUSAMMENFASSUNG
mRNA turnover mediated by the major protein-coding-region determinant of instability (mCRD) of the c-fos proto-oncogene transcript illustrates a functional interplay between mRNA turnover and translation. We show that the function of mCRD depends on its distance from the poly(A) tail. Five mCRD-associated proteins were identified: Unr, a purine-rich RNA binding protein; PABP, a poly(A) binding protein; PAIP-1, a poly(A) binding protein interacting protein; hnRNP D, an AU-rich element binding protein; and NSAP1, an hnRNP R-like protein. These proteins form a multiprotein complex. Overexpression of these proteins stabilized mCRD-containing mRNA by impeding deadenylation. We propose that a bridging complex forms between the poly(A) tail and the mCRD and ribosome transit disrupts or reorganizes the complex, leading to rapid RNA deadenylation and decay.