Merck
  • Replication study of 10 genes showing evidence for association with multiple sclerosis: validation of TMEM39A, IL12B and CBLB [correction of CLBL] genes.
Replication study of 10 genes showing evidence for association with multiple sclerosis: validation of TMEM39A, IL12B and CBLB [correction of CLBL] genes.
Multiple sclerosis (Houndmills, Basingstoke, England) (2011-12-24)
Jezabel Varadé, Manuel Comabella, Miguel A Ortiz, Rafael Arroyo, Oscar Fernández, M Jesús Pinto-Medel, María Fedetz, Guillermo Izquierdo, Miguel Lucas, Carlos López Gómez, Antonio Catalá Rabasa, Antonio Alcina, Fuencisla Matesanz, Iraide Alloza, Alfredo Antigüedad, María García-Barcina, David Otaegui, Javier Olascoaga, Albert Saiz, Yolanda Blanco, Xavier Montalbán, Koen Vandenbroeck, Elena Urcelay
ZUSAMMENFASSUNG
Ten genes previously showing different evidence of association with multiple sclerosis have been selected to validate. Eleven polymorphisms were genotyped with the iPLEX™ Sequenom in a well-powered collection of Spanish origin including 2863 multiple sclerosis cases and 2930 controls. Replication extended to the following polymorphisms: PKN2 (rs305217), GTF2B (rs7538427), EPHA4 (rs1517440), YTHDF3 (rs12115114), ANKFN1 (rs17758761) and PTPRM (rs4798571), which did not reach the threshold of significance in a follow-up of the first genome-wide association study (GWAS) conducted in multiple sclerosis; TMEM39A (rs1132200), which appeared as a newly identified susceptibility gene in the same study; a gene previously reaching GWAS significance in Italy, CBLB (rs9657904); IL12B (rs6887695, rs10045431), a susceptibility gene shared by diverse autoimmune diseases and, finally, another gene showing inconclusive association with multiple sclerosis, CNR1 (rs1049353). Pooled analysis corroborated the effect on MS predisposition of three genes: TMEM39A [rs1132200: p(M-H)=0.001; OR(M-H) (95% CI)= 0.84 (0.75-0.93)], IL12B [rs6887695: p(M-H)=0.03; OR(M-H) (95% CI)= 1.09 (1.01-1.17)] and CBLB [rs9657904: p(M-H)=0.01; OR(M-H) (95% CI)= 0.89 (0.81-0.97)].