Merck
  • Homodimerization of Ror2 tyrosine kinase receptor induces 14-3-3(beta) phosphorylation and promotes osteoblast differentiation and bone formation.
Homodimerization of Ror2 tyrosine kinase receptor induces 14-3-3(beta) phosphorylation and promotes osteoblast differentiation and bone formation.
Molecular endocrinology (Baltimore, Md.) (2007-08-25)
Yan Liu, John F Ross, Peter V N Bodine, Julia Billiard
ZUSAMMENFASSUNG
Ror2 receptor plays a key role in bone formation, but its signaling pathway is not completely understood. We demonstrate that Ror2 homodimerizes at the cell surface, and that dimerization can be induced by a bivalent antibody. Antibody-mediated dimerization causes receptor autophosphorylation and induces functional consequences of its signaling, including osteogenesis in mesenchymal stem cells and bone formation in organ culture. We further show that Ror2 associates with and phosphorylates 14-3-3beta scaffold protein. Endogenous Ror2 binds 14-3-3beta in U2OS osteosarcoma cells, and purified intracellular domain of Ror2 interacts with 14-3-3beta in vitro. 14-3-3beta Is tyrosine phosphorylated in U2OS cells, and this phosphorylation is inhibited by down-regulating Ror2 and enhanced by overexpressing the kinase. Purified Ror2 phosphorylates 14-3-3beta in vitro, confirming 14-3-3beta as the first identified Ror2 substrate. Down-regulating 14-3-3beta potentiates osteoblastogenesis in mesenchymal stem cells and increases bone formation in calvarial cultures, indicating that 14-3-3beta exerts a negative effect on osteogenesis. This raises a possibility that Ror2 induces osteogenic differentiation, at least in part, through a release of the 14-3-3beta-mediated inhibition. Our research forms a foundation for several new areas of investigation, including the molecular regulation of 14-3-3 by tyrosine phosphorylation and the role of this scaffold in osteogenesis.