Merck
  • Association of PS gene polymorphism and soluble P-selectin levels in atrial fibrillation thromboembolism population in Xinjiang.
Association of PS gene polymorphism and soluble P-selectin levels in atrial fibrillation thromboembolism population in Xinjiang.
Biochemical and biophysical research communications (2014-08-05)
Ling Bai, Zhiqiang Liu, Maerjiaen Bakeyi, Wuhong Lu, Pengyi He, Yuchun Yang, Muhuyati Wulasihan
ZUSAMMENFASSUNG
To investigate the association between the polymorphism of P choose element (p. selectin, PS) and soluble P-selectin levels in atrial fibrillation (AF) thromboembolism in Han and Uigur population of Xinjiang. Using ELISA method determination of plasma level of sPs. The frequency distributions of SNP sP-selectin gene promoter (-2123C/G) and SNP in exon region (Thr715Pro) were investigated by polymerase chain reaction (PCR)-restriction fragment length polymorphism and direct DNA sequence analysis among 302 Xinjiang Uigur and 340 age- and sex-matched Han people. Cases sPs exist significant difference serum level and the control group. The frequencies of the -2123C/G allele among the Uigur population had no significant differences from those of the Han population. Thr715Pro did not show any polymorphism in the two populations. The sP-selectin gene polymorphisms are associated with serum sP-selectin levels or thromboembolic events, suggesting that the patients with nonvalvular AF and thromboembolic events may have genetic susceptibility.