Merck
  • Unique expression features of cancer-type organic anion transporting polypeptide 1B3 mRNA expression in human colon and lung cancers.
Unique expression features of cancer-type organic anion transporting polypeptide 1B3 mRNA expression in human colon and lung cancers.
Clinical and translational medicine (2015-01-28)
Yuchen Sun, Tomomi Furihata, Seiya Ishii, Miki Nagai, Manami Harada, Osamu Shimozato, Takehiko Kamijo, Shinichiro Motohashi, Ichiro Yoshino, Atsuko Kamiichi, Kaoru Kobayashi, Kan Chiba
ZUSAMMENFASSUNG
We have previously identified the cancer-type organic anion transporting polypeptide 1B3 (Ct-OATP1B3) mRNA in several human colon and lung cancer tissues. Ct-OATP1B3 is a variant of the liver-type OATP1B3 (Lt-OATP1B3) mRNA, which is a hepatocyte plasma membrane transporter with broad substrate specificity. However, in cancer tissues, both the detailed characteristics of Ct-OATP1B3 mRNA expression and its biological functions remain unclear. With this point in mind, we sought to characterize Ct-OATP1B3 mRNA expression in colon and lung cancer tissues. In addition, we attempted to obtain functional implication of Ct-OATP1B3 in cancer cells. Matched pairs of cancer and normal tissues were collected from 39 colon cancer and 28 lung cancer patients. The OATP1B3 mRNA expression levels in each of these tissues were separately determined by quantitative real-time polymerase chain reaction. Mann-Whitney U test and Fisher's exact test were used in statistical analysis. The Ct-OATP1B3 functional expression in colon cancer cells was then examined by Western blotting and transport analyses. Ct-OATP1B3 mRNA, but not Lt-OATP1B3 mRNA, was abundantly expressed in colon cancer tissues at a higher detection frequency (87.2%) than that of the adjacent normal tissues (2.6%). Furthermore, it was found that Ct-OATP1B3 mRNA expression was often detected in early colon cancer stages (88.9%, n = 18), and that its expression was associated with well-differentiated colon cancer statuses. On the other hand, Ct-OATP1B3 mRNA also showed a predominant and cancer-associated expression profile in lung tissues, although at frequencies and expression levels that were lower than those obtained from colon cancer. As for attempts to clarify the Ct-OATP1B3 functions, neither protein expression nor transport activity could be observed in any of the cell lines examined. Based on the unique characteristics of the Ct-OATP1B3 mRNA expression profile identified in this study, Ct-OATP1B3 mRNA can be expected to become a biomarker candidate for use in colon (and lung) cancer diagnosis. Simultaneously, our results advance the possibility that Ct-OATP1B3 might play yet unidentified roles, in addition to transporter function, in cancer cell biology.