Merck
  • A multifunctional nanoprobe based on Au-Fe3O4 nanoparticles for multimodal and ultrasensitive detection of cancer cells.
A multifunctional nanoprobe based on Au-Fe3O4 nanoparticles for multimodal and ultrasensitive detection of cancer cells.
Chemical communications (Cambridge, England) (2013-04-23)
Jian Liu, Wei Zhang, Haoli Zhang, Zhengyin Yang, Tianrong Li, Baodui Wang, Xing Huo, Rui Wang, Haotai Chen
ZUSAMMENFASSUNG
A multifunctional nanoprobe, which can be used for dual modal imaging and the detection of cancer cells, has been reported.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(ethylenglycol)bis(amin), average Mn 3400