Merck
  • Alternative splicing for members of human mosaic domain superfamilies. I. The CH and LIM domains containing group of proteins.
Alternative splicing for members of human mosaic domain superfamilies. I. The CH and LIM domains containing group of proteins.
Molecular biology reports (2008-06-17)
Felix Friedberg
ZUSAMMENFASSUNG
In this paper we examine (restricted to homo sapiens) the products resulting from gene duplication and the subsequent alternative splicing for the members of a multidomain group of proteins which possess the evolutionary conserved calponin homology CH domain, i.e. an "actin binding domain", as a singlet and which, in addition, contain the conserved cysteine rich double Zn finger possessing Lim domain, also as a singlet. Seven genes, resulting from gene duplications, were identified that code for seven group members for which pre-mRNAs appear to have undergone multiple alternative splicing: Mical 1, 2 and 3 are located on chromosomes 6q21, 11p15 and 22q11, respectively. The LMO7 gene is present on chromosome 13q22 and the LIMCH1 gene on chromosome 4p13. Micall1 is mapped to chromosome 22q13 and Micall2 to chromosome 7p22. Translated Gen/Bank ESTs suggest the existence of multiple products alternatively spliced from the pre-mRNAs encoded by these genes. Characteristic indicators of such splicing among the proteins derived from one gene must include containment of some common extensive 100% identical regions. In some instances only one exon might be partly or completely eliminated. Sometimes alternative splicing is also associated with an increased frequency of creation of an exon or part of an exon from an intron. Not only coding regions for the body of the protein but also for its N- or -C ends could be affected by the splicing. If created forms are merely beginning at different starting points but remain identical in sequence thereafter, their existence as products of alternate splicing must be questioned. In the splicings, described in this paper, multiple isoforms rather than a single isoform appear as products during the gene expression.