Merck
3-(Pyridin-3-yl)propionic acid.
Acta crystallographica. Section E, Structure reports online (2010-01-01)
Andreas Lemmerer
ZUSAMMENFASSUNG
In the crystal of the title compound, C(8)H(9)NO(2), mol-ecules assemble to form C(8) chains along the b axis by N-H⋯O hydrogen bonds, supported by weaker C-H⋯O hydrogen bonded-inter-actions between adjacent chains.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Pyridinpropionsäure, 98%