Merck
  • Excited-state intramolecular proton transfer distinguishes microenvironments in single- and double-stranded DNA.
Excited-state intramolecular proton transfer distinguishes microenvironments in single- and double-stranded DNA.
The journal of physical chemistry. B (2008-09-05)
Andrey S Klymchenko, Volodymyr V Shvadchak, Dmytro A Yushchenko, Namrata Jain, Yves Mély
ZUSAMMENFASSUNG
Herein, the efficient interaction of an environment-sensitive fluorophore that undergoes excited-state intramolecular proton transfer (ESIPT) with DNA has been realized by conjugation of a 3-hydroxychromone (3HC) with polycationic spermine. On binding to a double-stranded DNA (dsDNA), the ratio of the two emission bands of the 3HC conjugates changes up to 16-fold, so that emission of the ESIPT product increases dramatically. This suggests an efficient screening of the 3HC fluorophore from the water molecules in the DNA complex, which is probably realized by its intercalation into dsDNA. In sharp contrast, the 3HC conjugates show only moderate changes in the dual emission on binding to a single-stranded DNA (ssDNA), indicating a much higher fluorophore exposure to water at the binding site. Thus, the 3-hydroxychromone fluorophore being conjugated to spermine discriminates the binding of this polycation to dsDNA from that to ssDNA. Consequently, ESIPT-based dyes are promising for monitoring the interaction of polycationic molecules with DNA and probing the microenvironment of their DNA binding sites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1′-Thiocarbonyldi-2(1H)-pyridon, 97%