Merck
Benzofuro[3,2-f][1]benzopyrans: a new class of antitubercular agents.
Bioorganic & medicinal chemistry (2006-04-18)
Soizic Prado, Hervé Ledeit, Sylvie Michel, Michel Koch, Jacques Christian Darbord, Stewart T Cole, François Tillequin, Priscille Brodin
ZUSAMMENFASSUNG
Alkylation of 2-hydroxydibenzofuran with 3-chloro-3-methyl-1-butyne, followed by Claisen rearrangement, gave access to 3,3-dimethyl-3Hbenzofuro[3,2-f][1]-benzopyran. Several derivatives modified at the pyran 1,2-double bond were prepared, including the corresponding dihydro compound and (+/-)-cis-diol, which was converted into diacetate and cyclic carbonate upon acylation. Both 3,3-dimethyl-3Hbenzofuro[3,2-f][1]benzopyran and 1,2-dihydro-3,3-dimethyl-3Hbenzofuro[3,2-f][1]benzopyran displayed significant activities when tested against Mycobacterium tuberculosis H37Rv and Beijing strains, with MIC99 in the range of 1-10 microg/ml. Further biological studies demonstrated good activities against drug-resistant mycobacterial strains. These compounds appear as promising specific antitubercular agents, since they exhibited neither significant cytotoxicity against mammal cells, nor effect on the growth of various bacteria and fungi.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Chlor-3-methyl-1-butin, 97%