Merck
  • Tirofiban as adjunctive therapy for acute coronary syndromes and percutaneous coronary intervention: a meta-analysis of randomized trials.
Tirofiban as adjunctive therapy for acute coronary syndromes and percutaneous coronary intervention: a meta-analysis of randomized trials.
European heart journal (2009-09-17)
Marco Valgimigli, Giuseppe Biondi-Zoccai, Matteo Tebaldi, Arnoud W J van't Hof, Gianluca Campo, Christian Hamm, Jurriën ten Berg, Leonardo Bolognese, Francesco Saia, Gian Battista Danzi, Carlo Briguori, Ertan Okmen, Spencer B King, David J Moliterno, Eric J Topol
ZUSAMMENFASSUNG
To perform a thorough and updated systematic review of randomized clinical trials comparing tirofiban vs. placebo or vs. abciximab. We searched for randomized trials comparing tirofiban vs. placebo or any active control. Odds ratios (OR) were computed from individual studies and pooled with random-effect methods. Thirty-one studies were identified involving 20,006 patients (12 874 comparing tirofiban vs. heparin plus placebo or bivalirudin alone, and 7132 vs. abciximab). When compared with placebo, tirofiban was associated at 30 days with a significant reduction in mortality [OR = 0.68 (0.54-0.86); P = 0.001] and death or myocardial infarction (MI) [OR = 0.69 (0.58-0.81); P < 0.001]. The treatment benefit persisted at follow-up but came at an increased risk of minor bleedings [OR = 1.42 (1.13, 1.79), P = 0.002] or thrombocytopenia. When compared with abciximab, mortality at 30 days did not differ [OR = 0.90 (0.53, 1.54); P = 0.70], but in the overall group tirofiban trended to increase the composite of death or MI [OR = 1.18 (0.96, 1.45); P = 0.11]. No such trend persisted at medium-term follow-up or when appraising studies testing tirofiban at 25 microg/kg bolus regimen. Tirofiban administration reduces mortality, the composite of death or MI and increases minor bleedings when compared with placebo. An early ischaemic hazard disfavouring tirofiban was noted when compared with abciximab in studies based on 10 but not 25 microg/kg tirofiban bolus regimen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tirofiban, ≥98.5% (HPLC)