Merck
  • Considerations in the development of an in vitro dissolution condition for lacidipine tablets: in vivo pharmacokinetic evaluation.
Considerations in the development of an in vitro dissolution condition for lacidipine tablets: in vivo pharmacokinetic evaluation.
Drug development and industrial pharmacy (2011-12-23)
Mingyu Sun, Jin Sun, Shuangfeng He, Yongjun Wang, Yinghua Sun, Xiaohong Liu, Zhonggui He
ZUSAMMENFASSUNG
In this study, a new discriminative dissolution condition for lacidipine tablets was developed by the established in vitro-in vivo relationship. Series of dissolution media of phosphate buffer solution (PBS) covering the pH range of 1-7.2 and pH 6.8 PBS containing different concentrations of sodium dodecyl sulfate (SDS), were prepared and used to investigate the dissolution behavior of lacidipine tablets. There was an obvious difference in the dissolution profiles of the both brands in pH 6.8 PBS medium containing 0.1% SDS. The pharmacokinetic study of the two lacidipine tablets was carried out in the healthy beagle dogs at a single dose of 4 mg. Statistical comparison of the AUC(₀₋₂₄), C(max), and T(max) showed a significant difference in the two brand tablets, coinciding with the dissolution performance with pH 6.8 PBS containing 0.1% SDS. The superiority of the proposed system, pH 6.8 PBS containing 0.1% SDS, could serve as a dissolution medium for lacidipine tablets, and more important it could discriminate the in vivo pharmacokinetic behavior for different brands of products. In summary, in vivo pharmacokinetic evaluation is essential to develop an appropriate in vitro dissolution condition for oral solid dosage forms of poorly soluble drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lacidipine, ≥98% (HPLC)