Merck
[Watch out for headaches at the end of a pregnancy! Do not miss an autoimmune hypositys].
Gynecologie, obstetrique & fertilite (2010-04-07)
F Lamazou, S Ouzounian, C Dupas, S Christin-Maitre
ZUSAMMENFASSUNG
We report the case of a 21-year-old pregnant women (36 WA) presenting intensive headaches and visual disturbances. Those sellar compression symptoms have made us suppose the diagnosis of autoimmune hypophysitis. MRI has confirmed the typical aspect of symmetric enlargement of the pituitary gland with a strong and homogenous enhancement after gadolinium. This pathology associated to a high morbidity and mortality due to partial or complete deficit of the anterior pituitary hormones, shall be reminded more often.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nefopam -hydrochlorid, ≥98% (HPLC)