Merck
[Chemical constituents from root of Prismatomeris tetrandra].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-02-14)
Jian-shuang Jiang, Zi-ming Feng, Pei-cheng Zhang
ZUSAMMENFASSUNG
To study the chemical constituents from root of Prismatomeris tetrandra. Compounds were isolated by chromatographic techniques on silica gel column. Their structures were elucidated by chemical and spectral methods. Six compounds were identified as 1-hydroxy-2-methylanthraquinone (I), 2-hydoxy-3-methoxyanthraquinone (II), 1, 3-dihydroxy-2-methoxyanthraquione (II), rubiadin (IV), rubiadin-1-methyl ether (V) and beta-sitosterol (V). I, II and III were isolated from the plant for the first time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Rubiadin, analytical standard