Merck
D and C Red No. 9: genotoxic or non-genotoxic carcinogen?
Mutation research (1992-03-01)
C Westmoreland, D Gatehouse
ZUSAMMENFASSUNG
The azo-compound, D and C Red No. 9 was assayed for genotoxicity in vivo using the rat micronucleus test and the rat ex vivo liver UDS assay. Uniformly negative results were obtained in both assays, even though large oral doses were used (2 g/kg). These results suggest that the tumorigenic effects of this compound in rats are mediated through non-genotoxic rather than a genotoxic mechanism. Further experiments using additional end-points such as 32P-post-labelling would further substantiate this conclusion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Pigment Rot 53, analytical standard