Merck
Altered visual function in monocarboxylate transporter 3 (Slc16a8) knockout mice.
American journal of physiology. Cell physiology (2008-06-06)
Lauren L Daniele, Brian Sauer, Shannon M Gallagher, Edward N Pugh, Nancy J Philp
ZUSAMMENFASSUNG
To meet the high-energy demands of photoreceptor cells, the outer retina metabolizes glucose through glycolytic and oxidative pathways, resulting in large-scale production of lactate and CO(2). Mct3, a proton-coupled monocarboxylate transporter, is critically positioned to facilitate transport of lactate and H(+) out of the retina and could therefore play a role in pH and ion homeostasis of the outer retina. Mct3 is preferentially expressed in the basolateral membrane of the retinal pigment epithelium and forms a heteromeric complex with the accessory protein CD147. To examine the physiological role of Mct3 in the retina, we generated mice with a targeted deletion in Mct3 (slc16A8). The overall retinal histology of 4- to 36-wk-old Mct3(-/-) mice appeared normal. In the absence of Mct3, expression of CD147 was lost from the basolateral but not apical RPE. The saturated a-wave amplitude (a(max)) of the scotopic electroretinogram (ERG) was reduced by approximately twofold in Mct3(-/-) mice relative to wild-type mice. A fourfold increase in lactate in the retina suggested a decrease in outer-retinal pH. In single-cell recordings from superfused retinal slices, saturating amplitudes of single rod photocurrents (J(max)) were comparable indicating that Mct3(-/-) mouse photoreceptor cells were inherently healthy. Based on these data, we hypothesize that disruption of Mct3 leads to a potentially reversible decrease in subretinal space pH, thereby reducing the magnitude of the light suppressible photoreceptor current.