Merck
  • Palladium-catalyzed carbopalladation and carbocyclization of arynes with aryl halides: a highly efficient route to functionalized triphenylenes.
Palladium-catalyzed carbopalladation and carbocyclization of arynes with aryl halides: a highly efficient route to functionalized triphenylenes.
Chemical communications (Cambridge, England) (2006-02-16)
Thiruvellore Thatai Jayanth, Chien-Hong Cheng
ZUSAMMENFASSUNG
Highly substituted triphenylene derivatives were prepared in good yields via the palladium-catalyzed carbocyclization of arynes with aryl iodides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Fluoriodbenzol, 99%