Merck
Alkylphenol biotransformations catalyzed by 4-ethylphenol methylenehydroxylase.
Applied and environmental microbiology (2003-06-06)
David J Hopper, Lisa Cottrell
ZUSAMMENFASSUNG
4-ethylphenol methylenehydroxylase from Pseudomonas putida JD1 acts by dehydrogenation of its substrate to give a quinone methide, which is then hydrated to an alcohol. It was shown to be active with a range of 4-alkylphenols as substrates. 4-n-propylphenol, 4-n-butylphenol, chavicol, and 4-hydroxydiphenylmethane were hydroxylated on the methylene group next to the benzene ring and produced the corresponding chiral alcohol as the major product. The alcohols 1-(4'-hydroxyphenyl)propanol and 1-(4'-hydroxyphenyl)-2-propen-1-ol, produced by the biotransformation of 4-n-propylphenol and chavicol, respectively, were shown to be R(+) enantiomers. 5-Indanol, 6-hydroxytetralin, 4-isopropylphenol, and cyclohexylphenol, with cyclic or branched alkyl groups, gave the corresponding vinyl compounds as their major products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Isopropyl-phenol, 98%