Merck
Effects of inhibitors of tubulin polymerization on GTP hydrolysis.
The Journal of biological chemistry (1981-09-10)
C M Lin, E Hamel
ZUSAMMENFASSUNG
The effects of a number of antimitotic drugs on the GTPase activity of tubulin were examined. The previously reported stimulation with colchicine and inhibition with podophyllotoxin and vinblastine wee confirmed. Maytansine, which competes with vinblastine in binding to tubulin, was comparable to the latter in inhibiting GTP hydrolysis. Nocodazole, which competes with colchicine in binding to tubulin, was significantly superior to colchicine in enhancing GTP hydrolysis. This superiority arose from the more rapid bindng of nocodazole to tubulin, as the two drugs had comparable activity when drug and tubulin were preincubated prior to the addition of GTP. Both colchicine and podophyllotoxin contain a trimethoxybenzene ring, while the closest structural analogy of nocodazole to colchicine includes the trimethoxybenzene ring. To explore this apparent paradox, we examined a number of simpler colchicine analogs for their effects on tubulin-dependent GTP hydrolysis. While tropolone was without effect, 3,4,5-trimethoxybenzaldehyde and 2,3,4-trimethoxybenzaldehyde stimulated the reaction. We therefore conclude that the trimethoxybenzene ring of colchicine is primarily responsible for the drug's stimulation of the GTPase activity of tubulin and that the inhibitory effect of podophyllotoxin must derive from the latter's tetrahydronaphthol moiety.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,3,4-Trimethoxybenzaldehyd, 99%