Merck
Total synthesis of (+/-)-halichlorine, (+/-)-pinnaic acid, and (+/-)-tauropinnaic acid.
Proceedings of the National Academy of Sciences of the United States of America (2004-08-10)
Hamish S Christie, Clayton H Heathcock
ZUSAMMENFASSUNG
The related marine natural products halichlorine, pinnaic acid, and tauropinnaic acid have been synthesized. The described route provided access to all three compounds from a common, late-stage intermediate. The synthesis began with 1-pyrrolidino-1-cyclopentene from which an intermediate possessing the three contiguous stereocenters of the natural products was synthesized in just four steps. Olefin cross metathesis followed by a hydrogenation/hydrogenolysis reaction stereoselectively formed the piperidine ring. Use of a beta-lactam group provided internal protection for the highly congested nitrogen atom during side-chain elaboration. The beta-lactam was subsequently reduced directly to an amino aldehyde, which after the Horner-Wadsworth-Emmons reaction was elaborated to pinnaic acid. The same amino aldehyde was also transformed into halichlorine after a thiol-mediated cyclization sequence to form the dehydroquinolizidine ring system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Pyrrolidino-1-cyclopenten, 98%