Merck
Proton-decoupled 19F spectroscopy of 5-FU catabolites in human liver.
Magnetic resonance in medicine (1997-03-01)
J Murphy-Boesch, C W Li, L He, K A Padavic-Shaller, W Negendank, T R Brown
ZUSAMMENFASSUNG
An RF network and a dual-tuned surface coil are described for obtaining proton-decoupled, NOE enhanced 19F spectra from a whole body clinical imager operating at 1.5 Tesia. The network removes 19F frequency noise from the decoupler transmitter, and prevents preamplifier saturation from high-level decoupling signals. Proton decoupling of 19F spectra was optimized using a sample of urine containing 5-fluorouracil (5-FU) and its catabolite fluoro-beta-alanine (FBAL). Proton-decoupled 19F spectroscopy in vivo is demonstrated by obtaining both nonlocalized spectra and spectra localized with three-dimensional chemical shift imaging from the liver of patients undergoing 5-FU chemotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Amino-2-fluoropropionic acid, 95%