Merck
  • Direct electron transfer from pseudoazurin to nitrous oxide reductase in catalytic N2O reduction.
Direct electron transfer from pseudoazurin to nitrous oxide reductase in catalytic N2O reduction.
Journal of inorganic biochemistry (2012-08-23)
Koyu Fujita, Mika Hirasawa-Fujita, Doreen E Brown, Yuji Obara, Fumihiro Ijima, Takamitsu Kohzuma, David M Dooley
ZUSAMMENFASSUNG
Pseudoazurin (PAz), a well-characterized blue copper electron-transfer protein, is shown herein to be capable of mediating electron transfer to the nitrous oxide reductase (N(2)OR) from Achromobacter cycloclastes (Ac). Spectroscopic measurements demonstrate that reduced PAz is efficiently re-oxidized by a catalytic amount of N(2)OR in the presence of N(2)O. Fits of the kinetics resulted in K(M) (N(2)O) and k(cat) values of 19.1±3.8 μM and 89.3±4.2s(-1) respectively. The K(M) (PAz) was 28.8±6.6 μM. The electrochemistry of Ac pseudoazurin (AcPAz) in the presence of Ac nitrous oxide reductase (AcN(2)OR) and N(2)O displayed an enhanced cathodic sigmoidal current-potential curve, in excellent agreement with the re-oxidation of reduced AcPAz during the catalytic reduction of N(2)O by AcN(2)OR. Modeling the structure of the AcPAz-AcN(2)OR electron transfer complex indicates that AcPAz binds near Cu(A) in AcN(2)OR, with parameters consistent with the formation of a transient, weakly-bound complex. Multiple, potentially efficient electron-transfer pathways between the blue-copper center in AcPAz and Cu(A) were also identified. Collectively, the data establish that PAz is capable of donating electrons to N(2)OR in N(2)O reduction and is a strong candidate for the physiological electron donor to N(2)OR in Ac.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Azurin aus Pseudomonas aeruginosa, from Pseudomonas aeruginosa, lyophilized powder