Merck
Stereoselective synthesis of β-(hydroxymethylaryl/alkyl)-α-methylene-γ-butyrolactones.
Organic letters (2011-04-15)
David M Hodgson, Eric P A Talbot, Barry P Clark
ZUSAMMENFASSUNG
Zinc or a chromium(II) source with 3-(bromomethyl)furan-2(5H)-one (3) and an aldehyde gives β-(hydroxymethylaryl/alkyl)-α-methylene-γ-butyrolactones 5 in good yields and high diastereoselectivities. The methodology is demonstrated in concise syntheses of (±)-hydroxymatairesinol (8) and (±)-methylenolactocin (10) by subsequent arylboronate conjugate addition and translactonization, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Methylen-γ-butyrolacton, 97%