Merck
  • Synthesis and cytostatic properties of daunorubicin derivatives, containing N-phenylthiourea or N-ethylthiourea moieties in the 3'-position.
Synthesis and cytostatic properties of daunorubicin derivatives, containing N-phenylthiourea or N-ethylthiourea moieties in the 3'-position.
The Journal of antibiotics (1991-02-01)
M N Preobrazhenskaya, E V Bakina, L S Povarov, E I Lazhko, L G Aleksandrova, J Balzarini, E De Clercq
ZUSAMMENFASSUNG
A series of phenylthiourea and ethylthiourea derivatives of daunorubicin and its congeners was prepared by reaction of the 3'-amino group of the antibiotic with phenylisothiocyanate or ethylisothiocyanate. S-Methylation yielded S-methylisothiouromium salts which when reacted with amines resulted in an intramolecular cyclization with the participation of the neighboring 4'-OH group. The structures and predominant conformations of the thiourea derivatives and daunorubicino(3'-N,4'-O-d)oxazolines were determined by 1H and 13C NMR. Cytostatic activities of the thiourea and oxazoline derivatives were compared with the cytostatic activities of N-methylurea and N-methyl-N-nitrosourea containing daunorubicin and its congeners. Carminomycin derivatives were endowed with the highest cytostatic activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Ethylthioharnstoff, 99%