Merck
The inhibitory effect of some new synthesized xanthates on mushroom tyrosinase activities.
Journal of enzyme inhibition and medicinal chemistry (2007-05-24)
M Alijanianzadeh, A A Saboury, H Mansuri-Torshizi, K Haghbeen, A A Moosavi-Movahedi
ZUSAMMENFASSUNG
Three iso-alkyldithiocarbonates (xanthates), as sodium salts, C3H7OCS2Na (I), C4H9OCS2Na (II) and C5H11OCS2Na (III), were synthesized, by the reaction between CS2 with the corresponding iso-alcohol in the presence of NaOH, and examined for inhibition of both cresolase and catecholase activities of mushroom tyrosinase (MT) from a commercial source of Agricus bisporus. 4-[(4-methylbenzo)azo]-1,2-benzendiol (MeBACat) and 4-[(4-methylphenyl)azo]-phenol (MePAPh) were used as synthetic substrates for the enzyme for the catecholase and cresolase reactions, respectively. Lineweaver-Burk plots showed different patterns of mixed and competitive inhibition for the three xanthates and also for cresolase and catecholase activities of MT. For cresolase activity, I and II showed a mixed inhibition pattern but III showed a competitive inhibition pattern. For catecholase activity, I showed mixed inhibition but II and III showed competitive inhibition. These new synthesized compounds are potent inhibitors of MT with K(i) values of 9.8, 7.2 and 6.1 microM for cresolase inhibitory activity, and also 12.9, 21.8 and 42.2 microM for catecholase inhibitory activity for I, II and III, respectively. They showed a greater inhibitory potency towards the cresolase activity of MT. Both substrate and inhibitor can be bound to the enzyme with negative cooperativity between the binding sites (alpha > 1) and this negative cooperativity increases with increasing length of the aliphatic tail in these compounds in both cresolase and catecholase activities. The cresolase inhibition is related to the chelating of the copper ions at the active site by a negative head group (S-) of the anion xanthate, which leads to similar values of K(i) for all three xanthates. Different K(i) values for catecholase inhibition are related to different interactions of the aliphatic chains of I, II and III with hydrophobic pockets in the active site of the enzyme.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
O-Isopropylxanthinsäure Kaliumsalz, 98%