Merck
Poly(N-acryloylsarcosine methyl ester) protein-resistant surfaces.
The journal of physical chemistry. B (2006-07-21)
D O H Teare, W C E Schofield, R P Garrod, J P S Badyal
ZUSAMMENFASSUNG
A new class of protein-resistant film based on N-substituted glycine derivatives is described. Pulsed plasma deposited poly(N-acryloylsarcosine methyl ester) coatings are shown to be resistant toward the adsorption of fibrinogen and lysozyme. Deposition and UV irradiation of the polymer through a masked grid are found to be effective ways for generating negative and positive image protein arrays, respectively, onto a range of different substrate materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sarcosin-methylester -hydrochlorid, ≥97.0% (T)