Merck
Role of histamine in the development of neurogenic inflammation of rat oral mucosa.
Agents and actions (1991-03-01)
A Györfi, A Fazekas, E Pósch, F Irmes, L Rosivall
ZUSAMMENFASSUNG
The mechanism of development of neurogenic inflammatory reaction induced by the topical application of capsaicin was studied in the oral mucosa of rats with or without histamine antagonist pretreatment. The existence of a cholinergic component of the vasodilation was investigated using a muscarinic receptor antagonist. Results indicated that the neurogenic inflammatory increases in vascular permeability and blood flow are mediated in part by H1-receptors, H2-receptors and cholinergic pathways are apparently not activated in these processes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Chlorpyramin -hydrochlorid, analytical standard, for drug analysis