- Determination of manganese in water samples by flame atomic absorption spectrometry after cloud point extraction.
Determination of manganese in water samples by flame atomic absorption spectrometry after cloud point extraction.
The Analyst (2001-05-09)
K C Teo, J Chen
PMID11340994
ZUSAMMENFASSUNG
Cloud point extraction has been used for the preconcentration of manganese, after the formation of a complex with 1-(2-thiazolylazo)-2-naphthol (TAN), and later analysis by flame atomic absorption spectrometry using octylphenoxypolyethoxyethanol (Triton X-114) as surfactant. The chemical variables affecting the separation phase and the viscosity affecting the detection process were optimized. Under the optimum conditions (i.e., pH = 9.2, [TAN] = 2.0 x 10(-5) mol l-1, [Triton X-114] = 0.05%, added methanol volume = 0.2 ml), preconcentration of 50 ml of sample solution permitted the detection of 0.28 ppb for manganese. The enhancement factor was 57.6. The proposed method has been applied to the determination of manganese in water samples.