Merck
  • Axin, a negative regulator of the wnt signaling pathway, directly interacts with adenomatous polyposis coli and regulates the stabilization of beta-catenin.
Axin, a negative regulator of the wnt signaling pathway, directly interacts with adenomatous polyposis coli and regulates the stabilization of beta-catenin.
The Journal of biological chemistry (1998-06-06)
S Kishida, H Yamamoto, S Ikeda, M Kishida, I Sakamoto, S Koyama, A Kikuchi
ZUSAMMENFASSUNG
The regulators of G protein signaling (RGS) domain of Axin, a negative regulator of the Wnt signaling pathway, made a complex with full-length adenomatous polyposis coli (APC) in COS, 293, and L cells but not with truncated APC in SW480 or DLD-1 cells. The RGS domain directly interacted with the region containing the 20-amino acid repeats but not with that containing the 15-amino acid repeats of APC, although both regions are known to bind to beta-catenin. In the region containing seven 20-amino acid repeats, the region containing the latter five repeats bound to the RGS domain of Axin. Axin and beta-catenin simultaneously interacted with APC. Furthermore, Axin stimulated the degradation of beta-catenin in COS cells. Taken together with our recent observations that Axin directly interacts with glycogen synthase kinase-3beta (GSK-3beta) and beta-catenin and that it promotes GSK-3beta-dependent phosphorylation of beta-catenin, these results suggest that Axin, APC, GSK-3beta, and beta-catenin make a tetrameric complex, resulting in the regulation of the stabilization of beta-catenin.