Merck
  • Relationship of in vitro minimum inhibitory concentrations of tilmicosin against Mannheimia haemolytica and Pasteurella multocida and in vivo tilmicosin treatment outcome among calves with signs of bovine respiratory disease.
Relationship of in vitro minimum inhibitory concentrations of tilmicosin against Mannheimia haemolytica and Pasteurella multocida and in vivo tilmicosin treatment outcome among calves with signs of bovine respiratory disease.
Journal of the American Veterinary Medical Association (2011-07-02)
David G McClary, Guy H Loneragan, Thomas R Shryock, Brandon L Carter, Carl A Guthrie, Marilyn J Corbin, Gerald D Mechor
ZUSAMMENFASSUNG
To determine associations between in vitro minimum inhibitory concentrations (MICs) of tilmicosin against Mannheimia haemolytica and Pasteurella multocida and in vivo tilmicosin treatment outcome among calves with clinical signs of bovine respiratory disease (BRD). Observational, retrospective, cohort study. 976 feeder calves with clinical signs of BRD enrolled in 16 randomized clinical trials. Records of clinical trials from October 26, 1996, to November 15, 2004, were searched to identify calves with BRD from which a single isolate of M haemolytica or P multocida was identified via culture of deep nasal swab samples prior to treatment with tilmicosin (10 mg/kg [4.5 mg/lb], SC) and for which MICs of tilmicosin against the isolate were determined. The MICs of tilmicosin against recovered isolates and response to tilmicosin treatment were evaluated. Tilmicosin resistance among M haemolytica and P multocida isolates was uncommon (6/745 [0.8%] and 16/231 [6.9%], respectively). Treatment outcome, defined as success or failure after tilmicosin treatment, did not vary with the MIC of tilmicosin against recovered isolates. The proportion of treatment failures attributed to M haemolytica isolates categorized as resistant (MIC of tilmicosin, ≥ 32 μg/mL) or not susceptible (MIC of tilmicosin, ≥ 16 μg/mL), was 0.2% and 0.5%, respectively. Recovery of tilmicosin-resistant M haemolytica or P multocida isolates was rare, and no association was detected between MIC of tilmicosin and treatment response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Tilmicosin, VETRANAL®, analytical standard, mix of isomers