Merck
  • Corticotropin-inhibiting peptide enhances aldosterone secretion by dispersed rat zona glomerulosa cells.
Corticotropin-inhibiting peptide enhances aldosterone secretion by dispersed rat zona glomerulosa cells.
The Journal of steroid biochemistry and molecular biology (1999-01-07)
L K Malendowicz, P Rebuffat, G G Nussdorfer, K W Nowak
ZUSAMMENFASSUNG
Corticotropin-inhibiting peptide (CIP), the 7-38 fragment of human ACTH(1-39), is known to act as an antagonist of ACTH receptors. Accordingly, CIP has been found to inhibit ACTH-stimulated glucocorticoid secretion of dispersed rat adrenocortical cells, without per se affecting the basal production. We confirmed these findings, but unexpectedly observed that CIP concentration-dependently raised basal aldosterone secretion from fresh suspensions of rat zona glomerulosa (ZG) cells, maximal effective concentration being 10(-6) M. CIP (10(-6) M) partially reversed the ZG-cell response to ACTH, but not to the Ca2+-dependent agonists angiotensin-II (ANG-II) and K+. The aspecific ANG-II-receptor antagonist saralasin (10(-6) M) blocked the aldosterone response of ZG cells to 10(-6) M CIP, and in the presence of the Ca2+-channel blocker verapamil CIP was ineffective. Collectively, these findings suggest that CIP enhances aldosterone secretion of rat ZG through a mechanism involving the activation of ANG-II receptors and the consequent rise in the cytosolic Ca2+ concentration. They also stress that this side-effect of CIP must be taken into account in interpreting the results of investigations on the adrenal cortex, where CIP has been employed as an ACTH-receptor antagonist.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 7-38 human, ≥97% (HPLC)