Merck
Determination of the diol content of chromatographic supports by capillary electrophoresis.
Journal of chromatography. A (1997-01-17)
A Chattopadhyay, D S Hage
ZUSAMMENFASSUNG
A capillary electrophoresis (CE) method was developed for determining the diol content of supports used in high-performance affinity or size exclusion chromatography. This method involved oxidizing the diol-bonded support with periodate, followed by the use of CE to separate and quantitate the iodate produced by this reaction. Both the oxidation and separation conditions were considered in optimizing this assay. The final method was performed by reacting a known amount of support with a 20-fold excess of periodate in pH 4.0, 0.5 M acetate buffer, with pyromellitic acid being used as an internal standard. After allowing 5-10 min for oxidation, the mixture was filtered and the filtrate was injected onto a 57 cm x 50 microns I.D. fused-silica capillary operated at 25 kV and containing pH 4.0, 0.5 M acetate as the running buffer. The total separation time was 5 min per run and gave a detection limit of 0.1 mM iodate (or 0.1 mumol diol groups) for a 6-nl injection of a 1-ml reaction mixture. By varying the amount the support that was assayed, this method could be used with either porous or non-porous supports. This technique showed good correlation with an iodometric titration but required much less sample and time to perform.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyromellitsäure, 96%