Merck
  • Analysis of steroids Part 50. Derivatization of ketosteroids for their separation and determination by capillary electrophoresis.
Analysis of steroids Part 50. Derivatization of ketosteroids for their separation and determination by capillary electrophoresis.
Journal of pharmaceutical and biomedical analysis (1996-06-01)
S Görög, M Gazdag, P Kemenes-Bakos
ZUSAMMENFASSUNG
4-Ene-3-ketosteroids and 17-ketosteroids were quantitatively transformed into the corresponding hydrazones using Girard P and T reagents, respectively. The positively charged derivatives were separated by capillary electrophoresis. The spectrophotometric characteristics of the derivatives permitted their sensitive detection in the 230-280 nm range. The steroids investigated included nortestosterone and its phenylpropionate, norethisterone and its oenanthate, d,l-norgestrel, dehydroepiandrosterone, androstenedione and ethisterone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Girard-Reagenz T, 98%