Merck
  • Effects of watercress consumption on metabolism of a tobacco-specific lung carcinogen in smokers.
Effects of watercress consumption on metabolism of a tobacco-specific lung carcinogen in smokers.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (1995-12-01)
S S Hecht, F L Chung, J P Richie, S A Akerkar, A Borukhova, L Skowronski, S G Carmella
ZUSAMMENFASSUNG
Epidemiological studies indicate that vegetable consumption protects against lung cancer in humans, but the protective constituents have not been identified. Phenethyl isothiocyanate (PEITC), which is release upon chewing of watercress (nasturtium officinale), is a chemopreventive agent against lung cancer induced by the tobacco-specific lung carcinogen 4- (methylnitrosamino)-1-(3-pyridyl-1-butanone (NNK) in rats and mice. PEITC inhibits the carcinogenicity of NNK by inhibiting its metabolic activation and thereby increasing the levels of detoxified metabolites excreted in urine. In this study, our goal was to determine whether watercress consumption would modify NNK metabolism in smokers. Eleven smokers maintained constant smoking habits and avoided cruciferous vegetables and other sources of isothiocyanates throughout the study. They donated 24-h urine samples on 3 consecutive days (baseline period). One to 3 days later, they consumed 2 ounces (56.8 g) of watercress at each meal for 3 days and donated 24-h urine samples on each of these days (watercress consumption period). One and 2 weeks later, they again donated 24-h urine samples on 2-3 consecutive days (follow-up periods). The samples were analyzed for two metabolites of NNK; 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol (NNAL) and [4-methylnitrosamino)-1-(3-pyridyl)but-1-yl]-beta-omega-D-glucosiduro nic acid (NNAL- Gluc). NNAL-Gluc is believed to be a detoxification product of NNK. The urine samples were also analyzed for PEITC-NAC, a metabolite of PEITC. Minimum exposure to PEITC during the watercress consumption period averaged 19-38 mg/day. Seven of the 11 subjects had increased levels of urinary NNAL plus NNAL-Gluc on days 2 and 3 of the watercress consumption period, compared to the baseline period. Overall, the increase in urinary NNAL plus NNAL-Gluc in this period was significant [mean +/- SD 0.924 +/- 1.12 nmol/24 h (33.5%); P < 0.01]. Urinary levels of NNAl plus NNAL-Gluc returned to near baseline levels in the follow-up periods. The percentage of increase in urinary NNAL plus NNAL-Gluc during days 2 and 3 of the watercress consumption period correlated with intake of PEITC during this period, as measured by total urinary PEITC-NAC (r = 0.62; P = 0.04). The results of this study support our hypothesis that PEITC inhibits this oxidative metabolism of NNK in humans, as seen in rodents, and support further development of PEITC as a chemopreventive agent against lung cancer. This is the first study to report an effect of vegetable consumption on metabolism of a lung carcinogen in humans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol, analytical standard