Merck
  • Combination of the chemotherapeutic agent 5-fluorouracil with an inhibitor of its catabolism results in increased micronucleus induction.
Combination of the chemotherapeutic agent 5-fluorouracil with an inhibitor of its catabolism results in increased micronucleus induction.
Biochemical and biophysical research communications (1994-09-15)
H Stopper, A Kühnel, B Podschun
ZUSAMMENFASSUNG
The rate limiting step in 5-fluorouracil catabolism is catalyzed by the enzyme dihydropyrimidine dehydrogenase. Since degradation of 5-fluorouracil decreases its efficacy in chemotherapy, the inhibition of its catabolism is a promising tool. We investigated the formation of micronuclei in vitro in mouse L5178Y cells. 5-fluorouracil induced an increase in micronucleus frequency, which could significantly be enhanced by the concurrent application of 2,6-dihydroxypyridine, an inhibitor of dihydropyrimidine dehydrogenase. The 5-fluorouracil concentration necessary to reach maximal genotoxic effects could be reduced to half in the presence of inhibitor 2,6-Dihydroxypyridine alone and the naturally occurring enzyme substrate uracil did not induce micronucleus formation. Combined application of the chemotherapeutic agent 5-fluorouracil and an inhibitor of its could reduce side-effects by lowering the effective dose of the active drug. With this study we provide further support for the usefulness of this concept.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,6-Dihydroxypyridin -hydrochlorid, 97%