Merck
Manganese, calcium, and saccharide binding to concanavalin A, as studied by ultrafiltration.
Archives of biochemistry and biophysics (1983-06-01)
J A Sophianopoulos, A J Sophianopoulos, W C MacMahon
ZUSAMMENFASSUNG
The binding of the ligands Mn2+, Ca2+, and methyl alpha-D-glucopyranoside to concanavalin A, purified as described (A.J. Sophianopoulos and J.A. Sophianopoulos (1981) Prep. Biochem. 11, 413-435), was studied by ultrafiltration in 0.2 M NaCl, pH 5.2 and pH 6.5 to 7, and at 23 to 25 degrees C. The association constant (Ka) of methyl alpha-D-glucopyranoside to concanavalin A was (2 +/- 0.2) X 10(3) M-1, both at pH 5.2 and 7. At pH 5.2 and in the absence of Ca2+, the Ka of Mn2+ to concanavalin A was (5 +/- 1) X 10(3) M-1, and in the presence of 1 mM Ca2+, the Ka was (9.1 +/- 2.1) X 10(5) M-1. At pH 6.5 Mn2+ bound to concanavalin A with a Ka of (7.3 +/- 1.8) X 10(5) M-1, and the binding affinity was virtually independent of the presence of Ca2+. Experiments of binding of 4-methylumbelliferyl alpha-D-mannopyranoside to concanavalin A indicated that at pH 5.2, binding of a single Mn2+ per concanavalin A monomer was sufficient to induce a fully active saccharide binding site. Ca2+ is not necessary for such activation, but rather it increases the affinity of concanavalin A for binding Mn2+.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Methylumbelliferyl α-D-Mannopyranosid, ≥97% (HPLC)