Merck
Preoperative skin antiseptics for preventing surgical wound infections after clean surgery.
The Cochrane database of systematic reviews (2013-04-02)
Jo C Dumville, Emma McFarlane, Peggy Edwards, Allyson Lipp, Alexandra Holmes
ZUSAMMENFASSUNG
Surgical site infection rates in the month following clean surgery vary from 0.6% (knee prosthesis) to 5% (limb amputation). Due to the large number of clean surgical procedures conducted annually the costs of these surgical site infections (SSIs) can be considerable in financial and social terms. Preoperative skin antisepsis using antiseptics is performed to reduce the risk of SSIs by removing soil and transient organisms from the skin where a surgical incision will be made. Antiseptics are thought to be toxic to bacteria and therefore aid their mechanical removal. The effectiveness of preoperative skin preparation is thought to be dependent on both the antiseptic used and the method of application, however, it is unclear whether preoperative skin antisepsis actually reduces postoperative wound infection, and, if so, which antiseptic is most effective. To determine whether preoperative skin antisepsis immediately prior to surgical incision for clean surgery prevents SSI and to determine the comparative effectiveness of alternative antiseptics. For this second update we searched the The Cochrane Wounds Group Specialised Register (searched 7 August 2012), The Cochrane Central Register of Controlled Trials (CENTRAL) (The Cochrane Library 2012, Issue 7), Ovid MEDLINE  (1950 to July Week 4 2012), Ovid MEDLINE (In-Process & Other Non-Indexed Citations August 06, 2012), Ovid EMBASE (1980 to 2012 Week 31), EBSCO CINAHL (2007 to 3 August 2012). Randomised controlled trials evaluating the use of preoperative skin antiseptics applied immediately prior to incision in clean surgery. There was no restriction on the inclusion of reports based on language of publication, date or publication status. Data extraction and assessment of risk of bias were undertaken independently by two review authors. Thirteen studies were included in this review (2,623 participants). These evaluated several different types of skin antiseptics - leading to 11 different comparisons being made. Although the antiseptics evaluated differed between studies, all trials involved some form of iodine. Iodine in alcohol was compared to alcohol alone in one trial; one trial compared povidone iodine paint (solution type not reported) with soap and alcohol. Six studies compared different types of iodine-containing products with each other and five compared iodine-containing products with chlorhexidine-containing products.There was evidence from one study suggesting that preoperative skin preparation with 0.5% chlorhexidine in methylated spirits led to a reduced risk of SSI compared with an alcohol based povidone iodine solution: RR 0.47 (95% CI 0.27 to 0.82). However, it is important to note that the trial does not report important details regarding the interventions (such as the concentration of povidone iodine paint used) and trial conduct, such that risk of bias was unclear.There were no other statistically significant differences in SSI rates in the other comparisons of skin antisepsis. Overall the risk of bias in included studies was unclear.A mixed treatment comparison meta-analysis was conducted and this suggested that alcohol-containing products had the highest probability of being effective - however, again the quality of this evidence was low. A comprehensive review of current evidence found some evidence that preoperative skin preparation with 0.5% chlorhexidine in methylated spirits was associated with lower rates of SSIs following clean surgery than alcohol-based povidone iodine paint. However this single study was poorly reported. Practitioners may therefore elect to consider other characteristics such as costs and potential side effects when choosing between alternatives.The design of future trials should be driven by the questions of high priority to decision makers. It may be that investment in at least one large trial (in terms of participants) is warranted in order to add definitive and hopefully conclusive data to the current evidence base. Ideally any future trial would evaluate the iodine-containing and chlorhexidine-containing solutions relevant to current practice as well as the type of solution used (alcohol vs. aqueous).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(vinylpyrrolidon)-Jod-Komplex