Merck
Induction of estradiol metabolism by dietary indole-3-carbinol in humans.
Journal of the National Cancer Institute (1990-06-06)
J J Michnovicz, H L Bradlow
ZUSAMMENFASSUNG
Dietary indoles in cruciferous vegetables induce cytochrome P450 enzymes and have prevented tumors in various animal models. Because estradiol metabolism is also cytochrome P450 mediated and linked to breast cancer risk, indoles may similarly reduce estrogen-responsive tumors in humans. We examined several indoles in female Sprague-Dawley rats for induction of hepatic estradiol 2-hydroxylation. The most potent inducer, indole-3-carbinol, was administered to humans (500 mg daily for 1 wk). It significantly increased the extent (mean +/- SEM) of estradiol 2-hydroxylation from 29.3% +/- 2.1% to 45.6% +/- 2.1% (P less than .001). These results indicate that indole-3-carbinol strongly influences estradiol metabolism in humans and may provide a new chemopreventive approach to estrogen-dependent diseases.