Merck
  • Combining BCR-ABL1 transcript levels at 3 and 6 months in chronic myeloid leukemia: implications for early intervention strategies.
Combining BCR-ABL1 transcript levels at 3 and 6 months in chronic myeloid leukemia: implications for early intervention strategies.
Blood (2013-02-06)
Pratap Neelakantan, Gareth Gerrard, Claire Lucas, Dragana Milojkovic, Philippa May, Lihui Wang, Christos Paliompeis, Marco Bua, Alistair Reid, Katayoun Rezvani, Stephen O'Brien, Richard Clark, John Goldman, David Marin
ZUSAMMENFASSUNG
Several groups have shown that that the BCR-ABL1 transcript level measured at 3 or 6 months after starting treatment with tyrosine kinase inhibitors strongly predicts clinical outcomes for patients with chronic myeloid leukemia. In this work, we asked whether the prognostic value of the 3-month transcript level could be improved by combining the 3- and 6-month results. We classified patients treated with imatinib and patients treated with dasatinib according to their transcript levels at 3 months and 6 months. The patients who met the 3-month landmark but failed the 6-month one had outcomes identical to those of patients who met both landmarks, whereas the patients who failed the first landmark but met the second one had prognoses similar to those who failed both landmarks. In summary, early intervention strategies can be based robustly just on the transcript level at 3 months. This trial was registered at www.clinicaltrials.gov as # NCT01460693.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Acebutolol -hydrochlorid, analytical standard
Acebutolol -hydrochlorid, European Pharmacopoeia (EP) Reference Standard