Merck
  • Venezuelines A-G, new phenoxazine-based alkaloids and aminophenols from Streptomyces venezuelae and the regulation of gene target Nur77.
Venezuelines A-G, new phenoxazine-based alkaloids and aminophenols from Streptomyces venezuelae and the regulation of gene target Nur77.
Bioorganic & medicinal chemistry letters (2012-11-21)
Jinwei Ren, Dong Liu, Li Tian, Yangye Wei, Peter Proksch, Jinzhang Zeng, Wenhan Lin
ZUSAMMENFASSUNG
Five new phenoxazine-based alkaloids venezuelines A-E (1-5) and two new aminophenols venezuelines F-G (6-7), as well as three known analogues exfoliazone, chandrananimycin D and carboxyexfoliazone were isolated from the fermentation broth of the marine-derived bacterium Streptomyces venezuelae. The structures of new compounds were determined on the basis of extensive spectroscopic analysis. The cytotoxic activity of these compounds against a panel of tumor cell lines were tested, while the regulation of gene target Nur77 of 2 and exfoliazone (8) were evaluated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenoxazin, 97%