Merck
Comparative proteomic analysis of drug sodium iron chlorophyllin addition to Hep 3B cell line.
The Analyst (2012-08-02)
Jun Zhang, Wenhai Wang, Fengying Yang, Xinwen Zhou, Hong Jin, Peng-yuan Yang
ZUSAMMENFASSUNG
The human hepatoma 3B cell line was chosen as an experimental model for in vitro test of drug screening. The drugs included chlorophyllin and its derivatives such as fluo-chlorophyllin, sodium copper chlorophyllin, and sodium iron chlorophyllin. The 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl-tetrazolium bromide (MTT) method was used in this study to obtain the primary screening results. The results showed that sodium iron chlorophyllin had the best LC(50) value. Proteomic analysis was then performed for further investigation of the effect of sodium iron chlorophyllin addition to the Hep 3B cell line. The proteins identified from a total protein extract of Hep 3B before and after the drug addition were compared by two-dimensional-gel-electrophoresis. Then 32 three-fold differentially expressed proteins were successfully identified by MALDI-TOF-TOF-MS. There are 29 unique proteins among those identified proteins. These proteins include proliferating cell nuclear antigen (PCNA), T-complex protein, heterogeneous nuclear protein, nucleophosmin, heat shock protein A5 (HspA5) and peroxiredoxin. HspA5 is one of the proteins which are involved in protecting cancer cells against stress-induced apoptosis in cultured cells, protecting them against apoptosis through various mechanisms. Peroxiredoxin has anti-oxidant function and is related to cell proliferation, and signal transduction. It can protect the oxidation of other proteins. Peroxiredoxin has a close relationship with cancer and can eventually become a disease biomarker. This might help to develop a novel treatment method for carcinoma cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chlorophyllin Natriumsalz