Merck
  • Novel amperometric assay for drug-DNA interaction based on an inhibitory effect on an electrocatalytic activity of DNA-Cu(II) complex.
Novel amperometric assay for drug-DNA interaction based on an inhibitory effect on an electrocatalytic activity of DNA-Cu(II) complex.
Biosensors & bioelectronics (2012-02-07)
Tingting Gu, Yasushi Hasebe
ZUSAMMENFASSUNG
A novel strategy of amperometric assay for drug-dsDNA interactions was developed based on an inhibitory effect of antimararial drug (quinacrine) on an electrocatalytic activity of DNA-Cu(II) complex. In this method, a DNA-Cu(II) complex immobilized DNA/polyallylamine(PAA) polyion complex membrane was used as a sensing element. The electrocatalytic activity of a DNA-Cu(II) complex for hydrogen peroxide reduction was reversibly inhibited by electron blocking effect of quinacrine-dsDNA interaction and this inhibitory effect was amplified by the hydrogen peroxide reduction. This phenomenon was utilized for development of a novel amperometric biosensor for DNA-binding drug. From the amperometric current-time curves, the response time of the sensor to 20 μM quinacrine was obtained about 20s, and the detection limit of the quinacrine was found to be 10 μM estimated to a signal-to-noise ratio of 3.0. Based on the change of steady-state catalytic current, the kinetic analysis of drug-dsDNA interaction can be done in a similar manner of enzyme inhibition, and the binding constant of the quinacrine with DNA can be calculated. This measurement method would be useful for screening of wide variety of DNA-binding drugs and highly toxic pollutants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Quinacrin -dihydrochlorid, ≥90%