Merck
  • Prostaglandin E2 inhibition of ketorolac 0.45%, bromfenac 0.09%, and nepafenac 0.1% in patients undergoing phacoemulsification.
Prostaglandin E2 inhibition of ketorolac 0.45%, bromfenac 0.09%, and nepafenac 0.1% in patients undergoing phacoemulsification.
Advances in therapy (2011-11-23)
Frank A Bucci, L David Waterbury
ZUSAMMENFASSUNG
We compared the prostaglandin E(2) (PGE(2)) inhibition of three topical nonsteroidal antiinflammatory drugs (NSAIDs): ketorolac 0.45%, bromfenac 0.09%, and nepafenac 0.1% at peak dosing levels in patients previously scheduled to undergo phacoemulsification. This was a single-center, double-masked observational study of 121 patients randomized to one of three NSAID treatment arms. Patients were instructed to take the NSAID per on-label dosing (twice daily [b.i.d.] for ketorolac 0.45% and bromfenac 0.09%, three times a day [t.i.d.] for nepafenac 0.1%) for 1 day before surgery, and were to instill one drop the morning of surgery. Each patient received an additional four doses 1 hour prior to undergoing phacoemulsification. After completion of the paracentesis site with a super blade, aqueous humor (0.15 mL) was collected through the peripheral clear cornea with a 30 G needle attached to a tuberculin (TB) syringe. Aqueous humor samples were stored at -40°C prior to analysis, and diluted 1:10 with diluent. Assays were conducted on multiple plates in duplicate (seven standards per plate). The mean (±SD) PGE(2) concentrations were 224.8±164.87 pg/mL for ketorolac 0.45% (n=42), 288.7±226.05 pg/mL for bromfenac 0.09% (n=41), and 320.4±205.6 pg/mL for nepafenac 0.1% (n=38). The difference between ketorolac 0.45% and nepafenac 0.1% was statistically significant (P=0.025). The difference between bromfenac 0.09% and nepafenac 0.1% was not significantly different (P=0.516). Ketorolac 0.45% achieved the greatest inhibition of PGE(2) compared to nepafenac 0.1% and bromfenac 0.09%. Ketorolac 0.45% may be more efficacious at controlling inflammation at the time of cataract surgery versus nepafenac 0.1% and bromfenac 0.09%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bromfenac sodium, ≥98% (HPLC)