Merck
  • Fucoxanthin, a marine carotenoid present in brown seaweeds and diatoms: metabolism and bioactivities relevant to human health.
Fucoxanthin, a marine carotenoid present in brown seaweeds and diatoms: metabolism and bioactivities relevant to human health.
Marine drugs (2011-11-11)
Juan Peng, Jian-Ping Yuan, Chou-Fei Wu, Jiang-Hai Wang
ZUSAMMENFASSUNG
The marine carotenoid fucoxanthin can be found in marine brown seaweeds, the macroalgae, and diatoms, the microalgae, and has remarkable biological properties. Numerous studies have shown that fucoxanthin has considerable potential and promising applications in human health. In this article, we review the current available scientific literature regarding the metabolism, safety, and bioactivities of fucoxanthin, including its antioxidant, anti-inflammatory, anticancer, anti-obese, antidiabetic, antiangiogenic and antimalarial activities, and its protective effects on the liver, blood vessels of the brain, bones, skin, and eyes. Although some studies have shown the bioavailability of fucoxanthin in brown seaweeds to be low in humans, many studies have suggested that a dietary combination of fucoxanthin and edible oil or lipid could increase the absorption rate of fucoxanthin, and thus it might be a promising marine drug.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fucoxanthin, carotenoid antioxidant
Supelco
Fucoxanthin, analytical standard