Merck
Discovery of a novel class of 2-aminopyrimidines as CDK1 and CDK2 inhibitors.
Bioorganic & medicinal chemistry letters (2011-06-21)
Jinho Lee, Kyoung-Hee Kim, Shinwu Jeong
ZUSAMMENFASSUNG
A series of new 2-(2-aminopyrimidin-4-yl)phenol derivatives were synthesized as potential antitumor compounds. Substitution with pyrrolidine-3,4-diol at the 4-position of phenol provided potent inhibitory activity against CDK1 and CDK2. X-ray crystal structural studies were performed to account for the effect of the substituent on both the enzymatic and cell growth inhibitory activities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminopyrimidin, 97%