Merck
  • Isoprenoid geranylgeranylacetone inhibits human colon cancer cells through induction of apoptosis and cell cycle arrest.
Isoprenoid geranylgeranylacetone inhibits human colon cancer cells through induction of apoptosis and cell cycle arrest.
Anti-cancer drugs (2010-08-21)
Naoyuki Yoshikawa, Nelson H Tsuno, Yurai Okaji, Kazushige Kawai, Yasutaka Shuno, Hirokazu Nagawa, Noriko Oshima, Koki Takahashi
ZUSAMMENFASSUNG
Geranylgeranylacetone (GGA), an isoprenoid compound, is a widely used antiulcer drug developed in Japan. GGA is structurally similar to plaunotol and geranylgeraniol, another isoprenoid reported to exert strong anticancer effects. In an earlier study, GGA was shown to inhibit ovarian cancer invasion by attenuating not only Rho activation, but also Ras-MAPK activation. In this study, we aimed to test whether GGA could have a therapeutic effect on colon cancer cells. As a result, we found that GGA induced a dose-dependent decrease in the proliferative activity through induction of cell apoptosis and cell cycle arrest in the G1 phase. The induction of apoptosis was mediated by the activation of both caspase-8 and caspase-9 pathways. The induction of G1 arrest was mediated by the increase of p21 and p27, and also the decrease of phosphorylated retinoblastoma protein levels. This study showed the potential anticancer activity of GGA. As this drug is already available in Japan for clinical use as an antiulcer/antigastritis agent, clinical trials will be designed to confirm its potential usefulness for cancer patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Geranylgeranylacetone